Human FOXP3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB877HU
MSPP-SEB877HUEA 665 USD
MSPP-SEB877HU
Human FOXP3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human FOXP3 (Forkhead Box P3). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.117 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Foxp3 is a member of the forkhead/winged-helix family of transcriptional regulators and functions as the master regulator in the development and function of regulatory T cells. Human FOXP3 genes contain 11 coding exons. Exon-intron boundaries are identical across the coding regions of the mouse and human genes. By genomic sequence analysis, the FOXP3 gene maps to chromosome Xp11.23. Foxp3 gene is mutated in the X-linked syndrome of immunodysregulation, polyendocrinopathy, and enteropathy (IPEX). These mutations were in the forkhead domain of FOXP3, indicating that the mutations may disrupt critical DNA interactions.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.