Human FETUB ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB860HU
MSPP-SEB860HUEA 665 USD
MSPP-SEB860HU
Human FETUB ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human FETUB (Fetuin B). The assay range is from 125 to 8000 pg/ml (Sandwich kit) with a sensitivity of 59 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
The fetuin family, which is part of the cystatin superfamily, encompasses a series of tightly related proteins that are synthesized mostly in the liver. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. Fetuin-B is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.