Human VEGF121 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB851HU
MSPP-SEB851HUEA 445 USD
MSPP-SEB851HU
Human VEGF121 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human VEGF121 (Vascular Endothelial Growth Factor 121). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.3 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
is a VEGF form that does not bind to neuropilins. Surprisingly, both np1 and neuropilin-2 (np2) enhanced VEGF121-induced phosphorylation of VEGFR-2 and VEGF121-induced proliferation of endothelial cells. The enhancement of VEGF121 activity by np1 was accompanied by a 10-fold increase in binding affinity to VEGFR-2 and was not associated with the formation of new VEGFR-2/np1 complexes. These observations suggest that neuropilins enhance the activity of VEGF forms that do not bind to neuropilins, indicate that np2 is a functional VEGF receptor, and imply that spontaneously formed VEGFR-2/np1 complexes suffice for efficient neuropilin mediated enhancement of VEGF activity.—Shraga-Heled, N. Kessler, O. Prahst, C. Kroll, J. Augustin, H. Neufeld, G. Neuropilin-1 and neuropilin-2 enhance VEGF121 stimulated signal transduction by the VEGFR-2 receptor.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.