Human Amya ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB836HU
MSPP-SEB836HUEA 665 USD
MSPP-SEB836HU
Human Amya ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human Amya (Amylase Alpha). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.55 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
α-Amylase is an enzyme that hydrolyses alpha-bonds of large alpha-linked polysaccharides such as starch and glycogen, yielding glucose and maltose. It is the major form of amylase found in humans and other mammals. It is also present in seeds containing starch as a food reserve, and is secreted by many fungi. The α-amylases are calcium metalloenzymes, completely unable to function in the absence of calcium. By acting at random locations along the starch chain, α-amylase breaks down long-chain carbohydrates, ultimately yielding maltotriose and maltose from amylose, or maltose, glucose and "limit dextrin" from amylopectin. Because it can act anywhere on the substrate, α-amylase tends to be faster-acting than β-amylase. In animals, it is a major digestive enzyme and its optimum pH is 6.7 to 7.0. In human physiology, both the salivary and pancreatic amylases are α-Amylases.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.