Rat NFkB ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB824RA
MSPP-SEB824RAEA 722 USD
MSPP-SEB824RA
Rat NFkB ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat NFkB (Nuclear Factor Kappa B). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.114 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Nuclear factor NF-kappa-B p105 subunit isa 105 kDa protein which can undergo cotranslational processing by the 26S proteasome to produce a 50 kDa protein. The 105 kDa protein is a Rel protein-specific transcription inhibitor and the 50 kDa protein is a DNA binding subunit of the NF-kappaB (NF-κB) protein complex.
NF-κB is a transcription factor that is activated by various intra- and extra-cellular stimuli such as cytokines, oxidant-free radicals, ultraviolet irradiation, and bacterial or viral products. Activated NF-κB translocates into the nucleus and stimulates the expression of genes involved in a wide variety of biological functions, over 200 known genes are targets of NF-κB in various cell types, under specific conditions.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.