Human SEPP1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB809HU
MSPP-SEB809HUEA 665 USD
MSPP-SEB809HU
Human SEPP1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human SEPP1 (Selenoprotein P1, Plasma). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.31 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Selenoprotein P is a selenoprotein containing multiple selenocysteine (Sec) residues, which are encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This selenoprotein is an extracellular glycoprotein, and is unusual in that it contains 10 Sec residues per polypeptide. It is a heparin-binding protein that appears to be associated with endothelial cells, and has been implicated to function as an antioxidant in the extracellular space. Several transcript variants, encoding either the same or different isoform, have been found for this gene.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.