Mouse S100A8 ELISA Kit
Supplier: 0000027876
SEB792MU
MSPP-SEB792MUEA 684 USD
MSPP-SEB792MU
Mouse S100A8 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse S100A8 (S100 Calcium Binding Protein A8). The assay range is from 7.8 to 500 pg/ml (Sandwich kit) with a sensitivity of 3.2 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
S100A8, a protein associated with a wide variety of inflammatory conditions, is heavily over-expressed in association with inflammation. Human S100A8 possesses neutrophil-repelling properties that result in an anti-inflammatory effect in vivo. The chemotactic activity of S100A8 on neutrophils was tested in Transwell chemotaxis assays. Analysis of the data indicates that S100A8 causes a repulsion of peripheral neutrophils, an activity that S100A8 loses upon its oxidation. S100A8 causes a strong anti-inflammatory effect in the rat air-pouch model of inflammation in vivo. These data highlight a naturally occurring novel anti-inflammatory pathway and provide potential molecular targets for the development of novel anti-inflammatory therapeutics.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.