Human IL26 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB695HU
MSPP-SEB695HUEA 665 USD
MSPP-SEB695HU
Human IL26 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL26 (Interleukin 26). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin-26 (IL-26), initially termed AK155, is a cellular sequence homolog to IL-10 belonging to the IL-10 cytokine family. Together with the related genes for interferon-gamma and IL-22/IL-TIF, il-26 maps to the human chromosomal region 12q15. The il-26 gene is one of the few differentially expressed genes specifying human T cells after growth-transformation with herpesvirus saimiri, a tumor virus of neo-tropical squirrel monkeys. Only herpesvirus saimiri-transformed T cells have been found to strongly over-express il-26 and to release the protein into the tissue culture supernatant. In a series of other T-cell lines and in native peripheral blood cells, il-26 is transcribed at low levels, but it is not detectable in B cells. Similarly to IL-10, the IL-26 protein forms homo-dimers. IL-26 is a candidate to contribute to the transformed phenotype of human T cells after infection by herpesvirus saimiri.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.