Human CXCL14 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB607HU
MSPP-SEB607HUEA 665 USD
MSPP-SEB607HU
Human CXCL14 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CXCL14 (Chemokine (C-X-C Motif) Ligand 14). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Chemokine (C-X-C motif) ligand 14 (CXCL14) is a small cytokine belonging to the CXC chemokine family that is also known as BRAK (for breast and kidney-expressed chemokine).Mature CXCL14 has many of the conserved features of the CXC chemokine subfamily but has some differences too, such as a shorter N-terminus and five extra amino acids in the region between its third and fourth cysteines. CXCL14 is constitutively expressed at high levels in many normal tissues, where its cellular source is thought to be fibroblasts.However, it is reduced or absent from most cancer cells.This chemokine is chemotactic for monocytes and can activate these cells in the presence of an inflammatory mediator called prostaglandin-E2 (PGE2).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.