Mouse SAP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB539MU
MSPP-SEB539MUEA 468 USD
MSPP-SEB539MU
Mouse SAP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse SAP (Serum Amyloid P Component). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.37 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
SAP can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. May also function as a calcium-dependent lectin. It is a precursor of amyloid component P which is found in basement membrane and associated with amyloid deposits.
SAP is a member of the pentraxins family, characterised by calcium dependent ligand binding and distinctive flattened beta-jellyroll structure similar to that of the legume lectins. The name 'pentraxin' is derived from the Greek word for five (penta) and berries (ragos) relating to the radial symmetry of five monomers forming a ring approximately 95 Aring across and 35 Aring deep. Human SAP has 51% sequence homology with C-reactive protein (CRP), a classical acute phase response plasma protein, and is a more distant relative to the 'long' pentraxins such as PTX3 (a cytockine modulated molecule) and several neuronal pentraxins. Both SAP and CRP are evolutionary conserved in all vertebrates and also found in distant invertebrates such as the horseshoe crab (Limulus polyphemus).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.