Human HBEGF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB479HU
MSPP-SEB479HUEA 528 USD
MSPP-SEB479HU
Human HBEGF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human HBEGF (Heparin Binding Epidermal Growth Factor Like Growth Factor). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Heparin-binding EGF-like growth factor (HB-EGF) is a member of the EGF family of proteins. It has been shown to play a role in wound healing, cardiac hypertrophy and heart development and function.
Diphtheria toxin inhibits protein synthesis in eukaryotic cells by catalyzing inactivation of elongation factor-2 (EF2). Entry of toxin into cells is dependent on binding to the cell surface.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.