General Species OVA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB459GE
MSPP-SEB459GEEA 449 USD
MSPP-SEB459GE
General Species OVA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species OVA (Ovalbumin). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.29 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Ovalbumin is the main protein found in egg white, making up 60 to 65% of the total protein. While Ovalbumin displays sequence and three-dimensional homology to the serpin superfamily, it is a noninhibitory serpin. While most serpins control such processes as fibrinolysis and coagulation by inhibiting serine proteases, the function of ovalbumin is unknown, although it is presumed to be a storage protein.
The ovalbumin protein of chickens is made up of 385 amino acids, and its relative molecular mass is 45 kDa. It is a glycoprotein with 4 sites of glycosylation.It is secreted from the cell. Unlike the signal sequences of most secreted proteins, Ovalbumin's signal sequence is not cleaved. Additionally it is not located directly on the N-terminus, but rather internally, starting at residue 21 and ending at residue 47.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.