Mouse TH ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB438MU
MSPP-SEB438MUEA 720 USD
MSPP-SEB438MU
Mouse TH ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse TH (Tyrosine Hydroxylase). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.56 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Tyrosine hydroxylase or tyrosine 3-monooxygenase is the enzyme responsible for catalyzing the conversion of the amino acid L-tyrosine to dihydroxyphenylalanine (DOPA). DOPA is a precursor for dopamine which in turn is a precursor for norepinephrine (noradrenaline) and epinephrine (adrenaline). In humans, tyrosine hydroxylase is encoded by the TH gene.The enzyme, an oxygenase, is found in the cytosol of all cells containing catecholamines. This initial reaction is the rate limiting step in the production of catecholamines.The enzyme is highly specific, not accepting indole derivatives - which is unusual as many other enzymes involved in the production of catecholamines do.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.