Human CD59 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB336HU
MSPP-SEB336HUEA 665 USD
MSPP-SEB336HU
Human CD59 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CD59 (Cluster of Differentiation 59). The assay range is from 12.5 to 800 pg/ml (Sandwich kit) with a sensitivity of 5.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Protectin, a cell surface glycoprotein that regulates complement-mediated cell lysis, and it is involved in lymphocyte signal transduction. This protein is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. This protein also plays a role in signal transduction pathways in the activation of T cells. Mutations in this gene cause CD59 deficiency, a disease resulting in hemolytic anemia and thrombosis, and which causes cerebral infarction. Multiple alternatively spliced transcript variants, which encode the same protein, have been identified for this gene.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.