Human PRF1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB317HU
MSPP-SEB317HUEA 665 USD
MSPP-SEB317HU
Human PRF1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PRF1 (Perforin 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.056 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Perforin is a cytolytic protein found in the granules of CD8 T-cells and NK cells. Perforin is a cytolytic protein found in the granules of Cytotoxic T lymphocytes (CTLs) and NK cells. Upon degranulation, perforin inserts itself into the target cell's plasma membrane, forming a pore. The lytic membrane-inserting part of perforin is the MACPF domain. This region shares homology with cholesterol-dependent cytolysins from Gram-positive bacteria. Perforin has structural and functional similarities to complement component 9 (C9). Like C9, this protein creates transmembrane tubules and is capable of lysing non-specifically a variety of target cells. This protein is one of the main cytolytic proteins of cytolytic granules, and it is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.