Human ITGaV ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB282HU
MSPP-SEB282HUEA 665 USD
MSPP-SEB282HU
Human ITGaV ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human ITGaV (Integrin Alpha V). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.057 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Integrin Alpha V encodes integrin alpha chain V. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. The I-domain containing integrin alpha V undergoes post-translational cleavage to yield disulfide-linked heavy and light chains, that combine with multiple integrin beta chains to form different integrins. Among the known associating beta chains (beta chains 1,3,5,6, and 8; 'ITGB1', 'ITGB3', 'ITGB5', 'ITGB6', and 'ITGB8'), each can interact with extracellular matrix ligands; the alpha V beta 3 integrin, perhaps the most studied of these, is referred to as the Vitronectin receptor (VNR). In addition to adhesion, many integrins are known to facilitate signal transduction.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.