Human IL31 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB179HU
MSPP-SEB179HUEA 665 USD
MSPP-SEB179HU
Human IL31 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL31 (Interleukin 31). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin-31 (IL-31) is a cytokine with a four-helix bundle structure, which is preferentially produced by type 2 helper T cells (Th2). The structure of IL-31 places it in the IL-6 family of cytokines. IL-31 signals via a receptor complex that is composed of IL-31 receptor A (IL-31RA) and oncostatin M receptor subunits. These receptor subunits are expressed in activated monocytes and in unstimulated epithelial cells. IL-31 is believed to play a role in inflammation of the skin. IL31, which is made principally by activated Th2-type T cells, interacts with a heterodimeric receptor consisting of IL31RA  and OSMR that is constitutively expressed on epithelial cells and keratinocytes. IL31 may be involved in the promotion of allergic skin disorders and in regulating other allergic diseases, such as asthma.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.