Human TFR ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB171HU
MSPP-SEB171HUEA 665 USD
MSPP-SEB171HU
Human TFR ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human TFR (Transferrin Receptor). The assay range is from 125 to 8000 pg/ml (Sandwich kit) with a sensitivity of 57 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Transferrin receptor  is a carrier protein for transferrin. It is needed for the import of iron into the cell and is regulated in response to intracellular iron concentration. Low iron concentrations promote increased levels of transferrin receptor, to increase iron intake into the cell. Thus, transferrin receptor maintains cellular iron homeostasis. Expression of human TFR1, but not human TFR2, in hamster cell lines markedly enhanced the infection of viruses pseudotyped with the glycoprotein of Machupo, Guanarito, and Junin viruses, but not with those of Lassa or lymphocytic choriomeningitis viruses. An anti-TFR1 antibody efficiently inhibited the replication of Machupo, Guanarito, Junin, and Sabia viruses, but not that of Lassa virus.TFR1 is a cellular receptor for New World hemorrhagic fever arenaviruses.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.