Human CR1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB123HU
MSPP-SEB123HUEA 665 USD
MSPP-SEB123HU
Human CR1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CR1 (Complement Receptor 1, Erythrocyte). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.061 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Erythrocyte complement receptor 1 is a member of the receptors of complement activation (RCA) family and is located in the 'cluster RCA' region of chromosome 1. The gene encodes a monomeric single-pass type I membrane glycoprotein found on erythrocytes, leukocytes, glomerular podocytes, and splenic follicular dendritic cells. The Knops blood group system is a system of antigens located on this protein. The protein mediates cellular binding to particles and immune complexes that have activated complement. Decreases in expression of this protein and/or mutations in its gene have been associated with gallbladder carcinomas, mesangiocapillary glomerulonephritis, systemic lupus erythematosus and sarcoidosis.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.