Human PIGR ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB074HU
MSPP-SEB074HUEA 665 USD
MSPP-SEB074HU
Human PIGR ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PIGR (Polymeric Immunoglobulin Receptor). The assay range is from 62.5 to 4000 pg/ml (Sandwich kit) with a sensitivity of 25.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Polymeric immunoglobulin receptor is a Fc receptor which facilitates the secretion of IgA and IgM. The poly-Ig receptor is expressed on several glandular epithelia including those of liver and breast. It mediates transcellular transport of polymeric immunoglobulin molecules. It is a member of the immunoglobulin superfamily (Hood et al. 1985). The receptor has 5 units with homology to the variable (V) units of immunoglobulins and a transmembrane region, which also has some homology to certain immunoglobulin variable regions.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.