Human VTN ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB041HU
MSPP-SEB041HUEA 665 USD
MSPP-SEB041HU
Human VTN ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human VTN (Vitronectin). The assay range is from 6.25 to 400 ng/ml (Sandwich kit) with a sensitivity of 2.74 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Vitronectin, is a 75 kDa glycoprotein in plasma and tissue. (S-protein is not to be confused with protein S.) A multifunctional protein, it promotes attachment and spreading of animal cells in vitro, inhibits cytolysis by the complement C5b-9 complex, and modulates antithrombin III-thrombin action in blood coagulation. The primary structure of vitronectin has been deduced from the sequence of its cloned cDNA. Polymorphism of vitronectin of plasma has been demonstrated. Sun and Mosher (1989) demonstrated that the frequencies are different in Orientals and Caucasians. By use of high resolution fluorescence in situ hybridization (FISH), Fink et al., (1992) mapped the VTN gene to 17q11. The localization was confirmed by cohybridization with a centromere-specific alphoid probe.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.