Mouse VIM ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB040MU
MSPP-SEB040MUEA 684 USD
MSPP-SEB040MU
Mouse VIM ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse VIM (Vimentin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.125 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Vimentin is a type III intermediate filament (IF) protein that is expressed in mesenchymal cells. IF proteins are found in all metazoan cells as well as bacteria. If, along with tubulin-based microtubules and actin-based microfilaments, comprise the cytoskeleton. All IF proteins are expressed in a highly developmentally-regulated fashion; vimentin is the major cytoskeletal component of mesenchymal cells. Because of this, vimentin is often used as a marker of mesenchymally-derived cells or cells undergoing an epithelial-to-mesenchymal transition (EMT) during both normal development and metastatic progression. A vimentin monomer, like all other intermediate filaments, has a central α-helical domain, capped on each end by non-helical amino (head) and carboxyl (tail) domains.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.