Mouse APP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB020MU
MSPP-SEB020MUEA 684 USD
MSPP-SEB020MU
Mouse APP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse APP (Amyloid Precursor Protein). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.112 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Amyloid precursor protein (APP) is an integral membrane protein expressed in many tissues and concentrated in the synapses of neurons. Its primary function is not known, though it has been implicated as a regulator of synapse formation and neural plasticity. APP is best known and most commonly studied as the precursor molecule whose proteolysis generates amyloid beta (Aβ), a 39- to 42-amino acid peptide whose amyloid fibrillar form is the primary component of amyloid plaques found in the brains of Alzheimer's disease patients.Several alternative splicing isoforms of APP have been observed in humans, ranging in length from 365 to 770 amino acids, with certain isoforms preferentially expressed in neurons; changes in the neuronal ratio of these isoforms have been associated with Alzheimer's disease.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.