Human a2M ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB017HU
MSPP-SEB017HUEA 560 USD
MSPP-SEB017HU
Human a2M ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human a2M (Alpha to 2-Macroglobulin). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.35 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Alpha2-macroglobulin is a major serum protein with diverse functions, including inhibition of protease activity and binding of growth factors, cytokines, and disease factors. It is also a panproteinase inhibitor that is found immunohistochemically in neuritic plaques. Alpha-2-macroglobulin is able to inactivate an enormous variety of proteinases (including serine-, cysteine-, aspartic- and metalloproteinases). Alpha-2-macroglobulin has in its structure a 35 aminoacid 'bait' region. Proteinases binding and cleaving the bait region become bound to alpha2M. The proteinase-α2M complex is recognised by macrophage receptors and cleared from the system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.