Mouse CYPA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA979MU
MSPP-SEA979MUEA 648 USD
MSPP-SEA979MU
Mouse CYPA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse CYPA (Cyclophilin A). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.64 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Cyclophilin A, also called Peptidyl-prolyl Isomerase A, PPIA, CYPA, and CYPH, was originally characterized for its ability to catalyze the transition between cis- and trans- proline residues critical for proper folding of proteins. Cyclophilin is also incorporated into many viruses, including HIV-1, where it has been speculated to be involved in functions such as viral assembly and infectivity. The immunosuppressive activity of cyclosporins has been correlated with their ability to form complexes with cyclophilins that inhibit calcineurin phosphatase activity and prevent incorporation of cyclophilin into viral particles. The cyclosporin/cyclophilin complex selectively binds and inactivates calcineurin, making it a useful inhibitor for studying calcineurin activity.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.