Human CP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA909HU
MSPP-SEA909HUEA 560 USD
MSPP-SEA909HU
Human CP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CP (Ceruloplasmin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.149 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Ceruloplasmin is is the major copper-carrying protein in the blood, and in addition plays a role in iron metabolism. It was first described in 1948.  It is an enzyme (EC 1.16.3.1) synthesized in the liver containing 6 atoms of copper in its structure. Ceruloplasmin carries 90% of the copper in plasma. The other 10% is carried by albumin, albumin may be confused at times to have a greater importance as a copper carrier because it binds copper less tightly than ceruloplasmin. Ceruloplasmin exhibits a copper-dependent oxidase activity, which is associated with possible oxidation of Fe₂ (ferrous iron) into Fe₃ (ferric iron), therefore assisting in its transport in the plasma in association with transferrin, which can only carry iron in the ferric state. The molecular weight of human ceruloplasmin is reported to be 151 kDa.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.