Human PPARg ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA886HU
MSPP-SEA886HUEA 630 USD
MSPP-SEA886HU
Human PPARg ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PPARg (Peroxisome Proliferator Activated Receptor Gamma). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.060 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Peroxisome Proliferator-Activated Receptor gamma is a member of the orphan nuclear receptor family. Oxidized metabolites of linoleic acid, 9-hydroxyctadienoic acid (9-HODE) and 13-HODE are activators and ligands of PPAR-gamma. PPAR-gamma; is expressed in white adipose tissue, intestinal mucosa, colon, spleen, monocytes, macrophages, retina, cartilage, osteoclasts, and skeletal muscle. PPAR-γ plays important roles in lipid and glucose metabolism, and has been implicated in obesity-related metabolic diseases such as hyperlipidemia, insulin resistance, and coronary artery disease. Three known family members are called PPAR alpha, delta, and gamma. Three N-terminal isoforms, called gamma1, gamma 2 and gamma 3, are known to arise by alternative splicing and promoter usage from the PPAR-gamma gene. RXR is an obligate partner for PPAR.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.