Bovine SAA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA885BO
MSPP-SEA885BOEA 756 USD
MSPP-SEA885BO
Bovine SAA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Bovine SAA (Serum Amyloid A). The assay range is from 6.25 to 400 ng/ml (Sandwich kit) with a sensitivity of 2.72 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Serum amyloid A protein is a protein encoded by the SAA1 gene. The serum amyloid A (SAA) protein is an acute phase apolipoprotein reactant produced mainly by hepatocytes and under regulation of inflammatory cytokines (summarized by Lundmark et al., 2002). The SAA cleavage product, designated amyloid protein A (AA), is deposited systemically as amyloid in vital organs including the liver, spleen, and kidneys in patients with chronic inflammatory diseases., There is very limited structural information about SAA because of its inherent poor solubility in the apolipoprotein form. It is intriguing to understand how such a small protein is able to mediate or directly carry out such a wide range of functions related to inflammatory reaction and other hostdefense mechanisms.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.