Human Lpa ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA842HU
MSPP-SEA842HUEA 630 USD
MSPP-SEA842HU
Human Lpa ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human Lpa (Lipoprotein, a). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.31 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Lipoprotein(a) consists of an LDL-like particle and the specific apolipoprotein(a), which is covalently bound to the apoB of the LDL like particle. Lp(a) plasma concentrations are highly heritable and mainly controlled by the apolipoprotein(a) gene located on chromosome 6q26-27. Apo(a) proteins vary in size due to a size polymorphism , which is caused by a variable number of so called kringle IV repeats in the LPA gene. This size variation at the gene level is expressed on the protein level as well, resulting in apo(a) proteins with 10 to >50 kringle IV repeats. These variable apo(a) sizes are known as 'apo(a) isoforms'. There is a general inverse correlation between the size of the apo(a)isoform and the Lp(a) plasma concentration which is caused by a variable rate of degradation before the apo(a) protein has matured for Lp(a) assembly.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.