Human CETP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA814HU
MSPP-SEA814HUEA 630 USD
MSPP-SEA814HU
Human CETP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CETP (Cholesteryl Ester Transfer Protein). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.29 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Cholesteryl ester transfer protein (CETP), also called plasma lipid transfer protein, is a plasma protein that facilitates the transport of cholesteryl esters and triglycerides between the lipoproteins. It collects triglycerides from very low-density (VLDL) or low-density lipoproteins (LDL) and exchanges them for cholesteryl esters from high-density lipoproteins (HDL), and vice versa. Most of the time, however, CETP does a homoexchange, trading a triglyceride for a triglyceride or a cholesteryl ester for a cholesteryl ester. Rare mutations leading to increased function of CETP have been linked to accelerated atherosclerosis. In contrast, a polymorphism (I405V) of the CETP gene leading to lower serum levels has also been linked to exceptional longevity
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.