Mouse ON ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA791MU
MSPP-SEA791MUEA 648 USD
MSPP-SEA791MU
Mouse ON ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse ON (Osteonectin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Osteonectin is a glycoprotein in the bone that binds calcium. It is secreted by osteoblasts during bone formation, initiating mineralization and promoting mineral crystal formation. Osteonectin also shows affinity for collagen in addition to bone mineral calcium. Some current research shows a correlation between osteonectin over expression and ampullary cancers and chronic pancreatitis. Osteonectin is an acidic, secreted extracellular matrix glycoprotein that plays a vital role in bone mineralization, cell-matrix interactions, and collagen binding. Osteonectin also increases the production and activity of matrix metalloproteinases, a function important to invading cancer cells within bone. Additional functions of osteonectin beneficial to tumor cells include angiogenesis, proliferation and migration.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.