Human PDL1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA788HU
MSPP-SEA788HUEA 630 USD
MSPP-SEA788HU
Human PDL1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PDL1 (Programmed Cell Death Protein 1 Ligand 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.056 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
CD274 (Cluster of Differentiation 274) also known as PD-L1 (Programed Death Ligand-1) or B7-H1 is a protein,PD-L1 binds to its receptor, PD-1, found on activated T cells, B cells, and myeloid cells, to modulate activation or inhibition. The affinity between PD-L1 and PD-1, as defined by the dissociation constant Kd, is 770 nM. Interestingly, PD-L1 also has an appreciable affinity for the costimulatory molecule CD80 (B7-1), but not CD86 (B7-2). CD80's affinity for PD-L1, 1.4 µM, is intermediate between its affinities for CD28 and CTLA-4 (4.0 µM and 400 nM, respectively). The related molecule PD-L2 has no such affinity for CD80 or CD86, but shares PD-1 as a receptor (with a stronger Kd of 140 nM).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.