Human PODXL ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA768HU
MSPP-SEA768HUEA 630 USD
MSPP-SEA768HU
Human PODXL ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PODXL (Podocalyxin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.054 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Podocalyxin belongs to the CD34/Podocalyxin family of sialomucins that share structural similarity and sequence homology. Podocalyxin is a major sialoprotein in the podocytes of the kidney glomerulus and is also expressed by both endothelium and multipotent hematopoietic progenitors. It has been identified as a novel cell surface marker for hemangioblasts, the common precursors of hematopoietic and endothelial cells.The  protein was originally identified as an important component of glomerular podocytes. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.