Human F1+2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA710HU
MSPP-SEA710HUEA 630 USD
MSPP-SEA710HU
Human F1+2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human F1+2 (Prothrombin Fragment 1+2). The assay range is from 3.9 to 250 pg/ml (Sandwich kit) with a sensitivity of 1.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Prothrombin is a carbohydrate-protein compound in plasma essential to coagulation. In response to bleeding, a complex series of clotting-factor interactions leads to its conversion by thromboplastin to thrombin, which transforms fibrinogen in plasma into fibrin. Fibrin and platelets combine to form a clot. Prothrombin and prothrombin fragment F1+2 (F1+2) were demonstrated in the tumor stroma on cancer cells and on small blood vessels in areas of neoangiogenesis at the host-tumor interface. F1+2 is an indicator of local activation of blood coagulation in cancer tissue. Thrombin itself is impossible to quantitate and so the use of surrogate markers is necessary. The measurement of F1+2 would be an excellent marker of thrombin generation. This is helped by the fact that F1+2 is not generated in vivo by any other mechanism. Fragment 1+2 has a half life of about 1 hour and is cleared from the bloodstream by the liver.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.