Human Hsp27 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA693HU
MSPP-SEA693HUEA 630 USD
MSPP-SEA693HU
Human Hsp27 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human Hsp27 (Heat Shock Protein 27). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.31 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Heat shock protein (HSP) 27 is one of the small HSPs. Like other small heat shock proteins, HSP27 is regulated at both the transcriptional and posttranslational levels. In response to stress, the expression level of HSP27 increases several-fold to confer cellular resistance to the adverse environmental change. HSP27 is phosphorylated at Ser15, Ser78 and Ser82 by MAPKAP kinase 2 as a result of the activation of the p38 MAP kinase pathway. Phosphorylation of HSP27 causes a change in its tertiary structure, which shifts from large homotypic multimers to dimers and monomers. It has been shown that phosphorylation and increased concentration of HSP27 modulates actin polymerization and reorganization.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.