Human PCT ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA689HU
MSPP-SEA689HUEA 560 USD
MSPP-SEA689HU
Human PCT ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PCT (Procalcitonin). The assay range is from 31.2 to 2,000 pg/ml (Sandwich kit) with a sensitivity of 12.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Procalcitonin (PCT) is a 116 amino acid residue peptide with molecular weight of about 13 kDa. PCT itself has no known hormonal activity. PCT belongs to a group of related proteins including calcitonin gene-related peptides I and II, amylin, adrenomodulin and calcitonin (CAPA peptide family). PCT, like other peptides of CAPA family, appears from the common precursor pre-procalcitonin consisting of 141 amino acids by removal of 25 amino acids from the N-terminus. PCT undergoes successive cleavages to form three molecules: N-terminal fragment (55 a.a.), calcitonin (32 a.a.) and katacalcin (21 a.a.). Under normal metabolic conditions, PCT is only present in the C cells of the thyroid gland. In bacterial infection and sepsis, however, intact PCT is found in the blood and, more importantly, its level is related to the severity of bacterial sepsis. Today, PCT is considered to be one of the earliest and most specific markers of sepsis.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.