Human FGF7 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA636HU
MSPP-SEA636HUEA 498 USD
MSPP-SEA636HU
Human FGF7 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human FGF7 (Fibroblast Growth Factor 7). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Keratinocyte growth factor is a protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is a potent epithelial cell-specific growth factor, whose mitogenic activity is predominantly exhibited in keratinocytes but not in fibroblasts and endothelial cells. Studies of mouse and rat homologs of this gene implicated roles in morphogenesis of epithelium, reepithelialization of wounds, hair development and early lung organogenesis.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.