Pig SELP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA569PO
MSPP-SEA569POEA 756 USD
MSPP-SEA569PO
Pig SELP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Pig SELP (P to Selectin). The assay range is from 6.25 to 400 ng/ml (Sandwich kit) with a sensitivity of 2.64 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
P-selectin is the largest of the known selectins at 140kDa. It contains nine consensus repeats (CR) and extends approximately 40 nm from the endothelial surface. P-selectin is expressed in a-granules of activated platelets and granules of endothelial cells. Within minutes of stimulation of the endothelial cells by inflammatory mediators such as histamine, thrombin, or phorbol esters, P-selectin is surface-expressed. Expression of P-selectin also occurs from the surgical trauma endured during preparation of the tissues for intravital microscopy. The expression is short-lived, reaching its peak after only ten minutes. Additional synthesis of P-selectin is brought about within two hours by cytokines such as interleukin-1 (IL-1) or tumor necrosis factor a (TNF-a).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.