Human ICAM1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA548HU
MSPP-SEA548HUEA 498 USD
MSPP-SEA548HU
Human ICAM1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human ICAM1 (Intercellular Adhesion Molecule 1). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 34 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
ICAM-1 is a type of intercellular adhesion molecule continuously present in low concentrations in the membranes of leukocytes and endothelial cells. Upon cytokine stimulation, the concentrations greatly increase. ICAM-1 can be induced by interleukin-1 (IL-1) and tumor necrosis factor alpha (TNFα) and is expressed by the vascular endothelium, macrophages, and lymphocytes. ICAM-1 is a ligand for LFA-1 (integrin), a receptor found on leukocytes.,When activated, leukocytes bind to endothelial cells via ICAM-1/LFA-1 and then transmigrate into tissues.ICAM-1 has been implicated in subarachnoid hemorrhage (SAH). Levels of ICAM-1 are shown to be significantly elevated in patients with SAH over control subjects in many studies.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.