Pig VCAM1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA547PO
MSPP-SEA547POEA 756 USD
MSPP-SEA547PO
Pig VCAM1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Pig VCAM1 (Vascular Cell Adhesion Molecule 1). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.55 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
VCAM-1 (CD106), a member of the immunoglobulin superfamily, is a cell surface protein expressed by activated endothelial cells and certain leukocytes (such as macrophages). VCAM-1 expression is induced by IL-1β, IL-4, TNF-α and IFN-γ. VCAM-1 binds to leukocyte integrins VLA-4 and α4β7. The rat and mouse SVCAM-1 proteins share approximately 76% amino acid similarity. VCAM-1 is a member of the Ig superfamily and encodes a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.