Pig IgA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA546PO
MSPP-SEA546POEA 756 USD
MSPP-SEA546PO
Pig IgA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Pig IgA (Immunoglobulin A). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 14.7 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Immunoglobulin A (IgA) is an antibody which plays a critical role in mucosal immunity. More IgA is produced in mucosal linings than all other types of antibody combined, between 3 and 5 g is secreted into the intestinal lumen each day. IgA has two subclasses (IgA1 and IgA2) and can exist in a dimeric form called secretory IgA (sIgA). In its secretory form, IgA is the main immunoglobulin found in mucous secretions, including tears, saliva, colostrum and secretions from the genito-urinary tract, gastrointestinal tract, prostate and respiratory epithelium. It is also found in small amounts in blood. The secretory component of sIgA protects the immunoglobulin from the being degraded by proteolytic enzymes, thus sIgA can survive in the harsh gastrointestinal tract environment and provide protection against microbes that multiply in body secretions.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.