Bovine IgM ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA543BO
MSPP-SEA543BOEA 588 USD
MSPP-SEA543BO
Bovine IgM ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Bovine IgM (Immunoglobulin M). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.71 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Immunoglobulin M, or IgM for short, is a basic antibody that is present on B cells. It is the primary antibody against A and B antigens on red blood cells. IgM is by far the physically largest antibody in the human circulatory system. It is produced after an animal has been exposed to an antigen for an extended time or when an animal is exposed to an antigen for the second time. IgM forms polymers where multiple immunoglobulins are covalently linked together with disulfide bonds, mostly as a pentamer but also as a hexamer. IgM has a molecular mass of approximately 900 kDa (in its pentamer form). Because each monomer has two antigen binding sites, a pentameric IgM has 10 binding sites. Typically, however, IgM cannot bind 10 antigens at the same time because the large size of most antigens hinders binding to nearby sites.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.