Mouse PAI1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA532MU
MSPP-SEA532MUEA 648 USD
MSPP-SEA532MU
Mouse PAI1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse PAI1 (Plasminogen Activator Inhibitor 1). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 12.3 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Plasminogen activator inhibitor-1 is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), the activators of plasminogen and hence fibrinolysis (the physiological breakdown of blood clots). It is a serine protease inhibitor (serpin) protein (SERPINE1). The other PAI, plasminogen activator inhibitor-2 (PAI-2) is secreted by the placenta and only present in significant amounts during pregnancy. In addition, protease nexin acts as an inhibitor of tPA and urokinase. PAI-1, however, is the main inhibitor of the plasminogen activators. PAI-1 is mainly produced by the endothelium (cells lining blood vessels), but is also secreted by other tissue types, such as adipose tissue.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.