Rat OxLDL ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA527RA
MSPP-SEA527RAEA 940 USD
MSPP-SEA527RA
Rat OxLDL ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat OxLDL (Oxidized Low Density Lipoprotein). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.9 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Hypercholesterolemia is closely associated with increased risk of atherosclerosis. The cholesterol accumulated in the atherosclerotic plaque is derived primarily from low-density lipoprotein (LDL). Oxidation of LDL is now accepted as a critical event in the atherogenic process. Free radicals like superoxide and nitric oxide (−O₂ , NO) generated in biological reactions in the body contribute to the oxidation of LDL. The NO radical has been shown to oxidize apolipoprotein B a constituant of LDL. Similarly, lipoxygenases and oxidants like peroxynitrite can oxidize the lipid moieties in LDL. It has been demonstrated that oxLDL and not native LDL is taken up by scavenger receptors on monocytes, smooth muscle cells and macrophages in the blood vessels. The oxidation of LDL increases the affinity of oxLDL to acetyl receptors of macrophages. As this pathway of oxLDL uptake is unregulated, it results in the formation of lipid-laden macrophages (foam cells).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.