Mouse FE ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA518MU
MSPP-SEA518MUEA 648 USD
MSPP-SEA518MU
Mouse FE ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse FE (Ferritin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.063 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Ferritin is an iron-protein complex formed from an intracellular acceptor called Apoferritin. Apoferritin is a large molecular weight 450000 protein produced by the liver. Iron as Fe (HO)3 linked to apoferritin is then stored in the cytoplasm of the reticuloendothelial system, liver, spleen and bone marrow. Ferritin is the body s iron storage protein functioning primarily as a site for iron storage from which iron may be mobilized in response to such stimuli a dietary change, blood loss or pregnancy. The direct quantitation of serum ferritin offers the physician a convenient and accurate measure of total body iron stores, by means of diagnosing iron-deficiency and anemia due to such causes as inflammation and hepatic or renal disease. In addition, serum ferritin concentration may be useful in detecting iron overload, which may allow the detection of idiopathic hemachromatosis in the precirrhotic storage.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.