Human CK5 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA488HU
MSPP-SEA488HUEA 630 USD
MSPP-SEA488HU
Human CK5 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CK5 (Cytokeratin 5). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.6 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic (i.e. different types of) keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. The type II cytokeratin genes are clustered in a region of chromosome 12q12-q13. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14.Eckert and Rorke (1988) cloned full-length KRT5 from a cultured keratinocyte cDNA library. The deduced 590-amino acid protein contains a central alpha-helical core region of about 310 amino acids with a 7-amino acid repeat found in coiled-coil structures. The N and C termini are serine rich and contain gly-gly-gly-x motifs.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.