Mouse NT-ProBNP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA485MU
MSPP-SEA485MUEA 680 USD
MSPP-SEA485MU
Mouse NT-ProBNP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse NT to ProBNP (N to Terminal Pro to Brain Natriuretic Peptide). The assay range is from 78 to 5,000 pg/ml (Sandwich kit) with a sensitivity of 28 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Brain-type Natriuretic Peptide (BNP) is a nonglycosylated peptide that is produced predominantly by ventricular myocytes and belongs to the natriuretic peptide family. Plasma NT-proBNP is a marker for congestive heart failure, while mature BNP (aa 103 - 134) promotes vasodilation and fluid and sodium excretion.BNP is co-secreted along with a 76 amino acid N-terminal fragment (NT-proBNP) which is biologically inactive. BNP binds to and activates the atrial natriuretic factor receptors NPRA, and to a lesser extent NPRB, in a fashion similar to atrial natriuretic peptide (ANP) but with 10-fold lower affinity. The biological half-life of BNP, however, is twice as long as that of ANP, and that of NT-proBNP is even longer, making these peptides better targets than ANP for diagnostic blood testing.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.