Human OC ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA471HU
MSPP-SEA471HUEA 630 USD
MSPP-SEA471HU
Human OC ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human OC (Osteocalcin). The assay range is from 0.625 to 40 ng/ml (Sandwich kit) with a sensitivity of 0.264 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Osteocalcin (OC) is the most abundant noncollagenous protein in mature bone where it constitutes 1 to 2% of the total protein. Synthesized by osteoblasts, it is incorporated into the bone matrix. Osteocalcin is a 49-residue protein with three gamma-carboxyglutamic acid residues, at positions 17, 21 and 24. These three residues confer on it a very strong ability to bind to hydroxyapatite. Osteocalcin is a noncollagenous protein found in bone and dentin. It is secreted by osteoblasts and thought to play a role in mineralization and calcium ion homeostasis. It has been stipulated that osteocalcin may also function as a negative regulator of bone formation, although its exact role is unknown.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.