Rabbit C3a ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA387RB
MSPP-SEA387RBEA 684 USD
MSPP-SEA387RB
Rabbit C3a ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rabbit C3a (Complement Component 3a). The assay range is from 7.8 to 500 ng/ml (Sandwich kit) with a sensitivity of 3.3 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
C3a is a one of the proteins formed by the cleavage of complement component 3; the other is C3b.C3a plays an important role in chemotaxis, though not as important a role as C5a.It is also an anaphylatoxin.Soluble C3-convertase, also known as C4b2a, catalyzes the proteolytic cleavage of C3 into C3a and C3b as part of the classical complement system as well as the mannan-binding lectin pathway. C3a is an anaphylotoxin, and C3b serves as an opsonizing agent. Factor I can cleave C3b into C3c and C3d, the latter of which plays a role in enhancing B cell responses.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.