Mouse AST ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA386MU
MSPP-SEA386MUEA 648 USD
MSPP-SEA386MU
Mouse AST ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse Lipoprotein lipase (LPL). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.61 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Lipoprotein lipase (EC 3.1.1.34) is an enzyme which hydrolyzes lipids in lipoproteins, like those found in chylomicrons and very low density lipoproteins (VLDL), into three free fatty acids and one glycerol molecule. It requires Apo-CII as a cofactor. Lipoprotein lipase is specifically found in endothelial cells lining the capillaries. Insulin is known to enhance LPL synthesis in adipocytes and its placement in the capillary endothelium. LPL has different isozymes in different tissues. The form that is in adipocytes is activated by insulin, whereas that in muscle and myocardium is not. This helps to explain why adipose cells gain fat in a well-fed state.A number sign is used with this entry because type I hyperlipoproteinemia can be caused by mutation in the lipoprotein lipase gene (LPL).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.